3-(Trifluorimetyyli)fenyyliboronihappo 97+%

Toimittaja: Thermo Fisher Scientific
Varoitus

359750010 359750050
ACRO359750010EA 34.3 EUR
ACRO359750010 ACRO359750050
3-(Trifluorimetyyli)fenyyliboronihappo 97+%
3-(Trifluorimetyyli)fenyyliboronihappo
Kaava: C₇H₆BF₃O₂
Sulamispiste: 160…165 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00151854
CAS-numero: 1423-26-3

Order Now

Testitulosten erittely

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 161 to 167 °C
HPLC ≥97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR