3-Pyridineboronic acid 95%

Toimittaja: Thermo Fisher Scientific
Varoitus

359650010 359650050
ACRO359650010EA 92.8 EUR
ACRO359650010 ACRO359650050
3-Pyridineboronic acid 95%
3-Pyridineboronic acid
Kaava: C₅H₆BNO₂
Sulamispiste: >300 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00674177
CAS-numero: 1692-25-7

Order Now

Testitulosten erittely

Appearance White to yellow to light orange Powder or crystals and/or chunks
Infrared spectrum Conforms
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR