4-Phenoxyphenylboronic acid 95+%

Toimittaja: Thermo Fisher Scientific
Varoitus

359640010 359640050 359640250
ACRO359640010EA 41.7 EUR
ACRO359640010 ACRO359640050 ACRO359640250
4-Phenoxyphenylboronic acid 95+%
4-Phenoxyphenylboronic acid
Kaava: C₁₂H₁₁BO₃
Sulamispiste: 141…145 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00093312
CAS-numero: 51067-38-0

Order Now

Testitulosten erittely

Appearance White to off-white Powder and/or chunks
Infrared spectrum Conforms
HPLC ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR