1-Ethyl-3-methylimidazolium bromide 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

356670250 356670050 356671000
ACRO356670250EA 67.3 EUR
ACRO356670250 ACRO356670050 ACRO356671000
1-Ethyl-3-methylimidazolium bromide 98%
1-Ethyl-3-methylimidazolium bromide
Kaava: C₆H₁₁BrN₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD03427610
CAS-numero: 65039-08-9

Order Now

Testitulosten erittely

Appearance Yellow Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR