3-Klooribentseenisulfonyylikloridi 98%

Toimittaja: Thermo Fisher Scientific
Vaara

356480010 356480050 356480250
ACRO356480010EA 28.3 EUR
ACRO356480010 ACRO356480050 ACRO356480250
3-Klooribentseenisulfonyylikloridi 98%
3-Klooribentseenisulfonyylikloridi
Kaava: C₆H₄Cl₂O₂S
Kiehumispiste: 102 °C (,9 mmHg)
Tiheys: 1,499 g/cm³
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00051697
CAS-numero: 2888-06-4
EINECS: 220-753-0
UN: 3265
ADR: 8,II

Order Now

Testitulosten erittely

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5670 to 1.5690 (20°C, 589 nm)
Titration RCOCl ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR