Thiophene-2-ethylamine 98%

Toimittaja: Thermo Fisher Scientific
Vaara

354270010
ACRO354270010EA 40.9 EUR
ACRO354270010
Thiophene-2-ethylamine 98%
Thiophene-2-ethylamine
Kaava: C₆H₉NS
Kiehumispiste: 103…105 °C (15 mmHg)
Storage Temperature: Jääkaappi
MDL numero: MFCD00051495
CAS-numero: 30433-91-1
EINECS: 250-196-9
UN: 2735
ADR: 8,III

Order Now

Testitulosten erittely

Appearance Colorless to yellow to orange Liquid
Infrared spectrum Conforms
Refractive index 1.5500 to 1.5540 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR