Potassium tetraoxalate dihydrate 99%, extra pure

Toimittaja: Thermo Fisher Scientific
Varoitus

352111000 352110010 352115000
ACRO352111000EA 33.3 EUR
ACRO352111000 ACRO352110010 ACRO352115000
Potassium tetraoxalate dihydrate 99%, extra pure
Potassium tetraoxalate dihydrate
Kaava: C₄H₃KO₈·2H₂O
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00150443
CAS-numero: 6100-20-5
EINECS: 204-874-6
ADR: 6.1,III

Order Now

Testitulosten erittely

Appearance Colorless to white Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying 12 % to 16 % (1 g, 105°C)
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR