Metyyli-3-hydroksibentsoaatti 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

352000250 352001000
ACRO352000250EA 62.7 EUR
ACRO352000250 ACRO352001000
Metyyli-3-hydroksibentsoaatti 98%
Metyyli-3-hydroksibentsoaatti
Kaava: C₈H₈O₃
Kiehumispiste: 280…281 °C (709 mmHg)
Sulamispiste: 70…74 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002295
CAS-numero: 19438-10-9
EINECS: 243-071-5

Order Now

Testitulosten erittely

Appearance White to beige Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 70 to 74 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR