(±)-1-fenyylietanoli 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: (±)-sec-Phenethyl alcohol, Styrene alcohol, (±)-α-Methylbenzyl alcohol, (±)-1-Phenylethyl alcohol, 1-Phenylethanol, Methyl phenyl carbinol, α-Methylbenzyl alcohol, Styrallyl alcohol

351881000 351885000
ACRO351881000EA 37 EUR
ACRO351881000 ACRO351885000
(±)-1-fenyylietanoli 97%
(±)-1-fenyylietanoli
Kaava: C₆H₅CH(OH)CH₃
Kiehumispiste: 401 °C (1013 hPa)
Sulamispiste: 20 °C
Tiheys: 1,01 g/cm³
Leimahduspiste: 86 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00004508
CAS-numero: 98-85-1
EINECS: 202-707-1
UN: 2937
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa955c5829772d9b27ccf96aa5e9b66b661e94a5&vid=43f0c1e8ef3c916809a95af92edf59ec33769eab&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5250 to 1.5290 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR