Kloroformi-D1 99.8 atom % D stabiloitu for NMR spectroscopy

Toimittaja: Thermo Fisher Scientific
Vaara


Stabilization: Stabilised with silver

351420250 351421000 351425000
ACRO351420250EA 54.7 EUR
ACRO351420250 ACRO351421000 ACRO351425000
Kloroformi-D1 99.8 atom % D stabiloitu for NMR spectroscopy
Kloroformi-D1
Kaava: CDCl₃
Kiehumispiste: 60,8 °C (1013 hPa)
Sulamispiste: –64 °C
Tiheys: 1,5 g/cm³ (20 °C)
Storage Temperature: Jääkaappi
MDL numero: MFCD00000827
CAS-numero: 865-49-6
EINECS: 212-742-4
UN: 1888
ADR: 6.1,III

Order Now

Testitulosten erittely

Appearance Clear colorless Liquid
Water ≤0.02 % (Coulometric)
D-Enrichment ≥99.75 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR