Asetoni-D6 99.9 atom % D + 0.03% (v/v) TMS for NMR spectroscopy

Toimittaja: Thermo Fisher Scientific
Vaara

351380100 351380500
ACRO351380100EA 62.1 EUR
ACRO351380100 ACRO351380500
Asetoni-D6 99.9 atom % D + 0.03% (v/v) TMS for NMR spectroscopy
Asetoni-D6
Kaava: D₃CC(O)CD₃
Kiehumispiste: 55,5 °C (1013 hPa)
Sulamispiste: –93 °C
Tiheys: 0,87 g/cm³
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00044635
CAS-numero: 666-52-4
EINECS: 211-563-9
UN: 1090
ADR: 3,II

Order Now

Testitulosten erittely

Appearance Clear colorless Liquid
Water ≤0.03 % (Coulometric)
D-Enrichment ≥99.85 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR