(2-Bromifenyyli)asetoni 99%

Toimittaja: Thermo Fisher Scientific

350590010 350590050
ACRO350590010EA 16.5 EUR
ACRO350590010 ACRO350590050
(2-Bromifenyyli)asetoni 99%
(2-Bromifenyyli)asetoni
Kaava: C₉H₉BrO
Storage Temperature: Huoneenlämpö
MDL numero: MFCD03410431
CAS-numero: 21906-31-0

Order Now

Testitulosten erittely

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5575 to 1.5595 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR