3-Bromophenethylamine 99%

Toimittaja: Thermo Fisher Scientific
Vaara

350580010 350580050 350580250
ACRO350580010EA 50.8 EUR
ACRO350580010 ACRO350580050 ACRO350580250
3-Bromophenethylamine 99%
3-Bromophenethylamine
Kaava: C₈H₁₀BrN
Tiheys: 1,406 g/cm³
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01310790
CAS-numero: 58971-11-2
UN: 2735
ADR: 8,III

Order Now

Testitulosten erittely

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5735 to 1.5755 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR