Triethylammonium bromide 99%

Toimittaja: Thermo Fisher Scientific
Varoitus

349721000 349725000
ACRO349721000EA 42.1 EUR
ACRO349721000 ACRO349725000
Triethylammonium bromide 99%
Triethylammonium bromide
Kaava: C₆H₁₅N·HBr
Leimahduspiste: >65 °C
MDL numero: MFCD00054287
CAS-numero: 636-70-4
EINECS: 211-263-8

Order Now

Testitulosten erittely

Appearance White to off-white Crystalline powder or flakes
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR