Brij® 98

Toimittaja: Thermo Fisher Scientific
Varoitus

347185000 347181000
ACRO347185000EA 102 EUR
ACRO347185000 ACRO347181000
Brij® 98
Brij® 98
Kaava: C₅₈H₁₁₆O₂₁
Kiehumispiste: >100 °C (760 mmHg)
Sulamispiste: 30 °C
Tiheys: 1,07 g/cm³
Leimahduspiste: >150 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00074369
CAS-numero: 9004-98-2
EINECS: 500-016-2

Order Now

Testitulosten erittely

Appearance White to cream Waxy solid
Infrared spectrum Conforms
Water ≤3 %
Additional info remark: without antioxidant
Impurity ≤1 ppm (Free ethyleneoxide)
Dioxane ≤10 ppm
Acid value ≤1 mg KOH/g
Hydroxyl value 50 to 65 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR