2,4-Hexadiene mixture of isomers 90% tech.

Toimittaja: Thermo Fisher Scientific
Vaara

346670010 346670050
ACRO346670010EA 47.1 EUR
ACRO346670010 ACRO346670050
2,4-Hexadiene mixture of isomers 90% tech.
2,4-Hexadiene
Kaava: C₆H₁₀
Kiehumispiste: 82 °C (1013 hPa)
Sulamispiste: –79 °C
Tiheys: 0,72 g/cm³
Leimahduspiste: –8 °C
MDL numero: MFCD00009297
CAS-numero: 592-46-1
EINECS: 209-757-3
UN: 2458
ADR: 3,II

Order Now

Testitulosten erittely

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
GC ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR