Methylboronic acid 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

344670010 344670050
ACRO344670010EA 56.2 EUR
ACRO344670010 ACRO344670050
Methylboronic acid 97%
Methylboronic acid
Kaava: CH₅BO₂
Sulamispiste: 88…90 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00002105
CAS-numero: 13061-96-6

Order Now

Testitulosten erittely

Appearance White to beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 87 to 91 °C
Titration with NaOH 96.0 to 104.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR