Piperonyl butoxide 90% tech.

Toimittaja: Thermo Fisher Scientific
Varoitus

334161000 334165000
ACRO334161000EA 66.3 EUR
ACRO334161000 ACRO334165000
Piperonyl butoxide 90% tech.
Piperonyl butoxide
Kaava: C₁₉H₃₀O₅
Kiehumispiste: 180 °C (1 mmHg)
Leimahduspiste: 170 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00005842
CAS-numero: 51-03-6
UN: 3082
ADR: 9,III

Order Now

Testitulosten erittely

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4970 to 1.5070 (20°C, 589 nm)
GC ≥88 %
Specific gravity (20°C) 1.055 to 1.065


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR