Antimoni(V)sulfidi, pure

Toimittaja: Thermo Fisher Scientific
Vaara

330410010 330410500 330412500
ACRO330410010EA 240 EUR
ACRO330410010 ACRO330410500 ACRO330412500
Antimoni(V)sulfidi, pure
Antimoni(V)sulfidi
Kaava: S₅Sb₂
Tiheys: 4,12 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00016321
CAS-numero: 1315-04-4
EINECS: 215-255-5
UN: 3179
ADR: 4.1,II
Merck Index: 12,00738

Order Now

Testitulosten erittely

Appearance Yellow to orange-red Powder
Assay ≥54 % (Sb)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR