1-(4-Chlorophenyl)ethanamine 90%

Toimittaja: Thermo Fisher Scientific
Vaara

329470050 329470010
ACRO329470050EA 164 EUR
ACRO329470050 ACRO329470010
1-(4-Chlorophenyl)ethanamine 90%
1-(4-Chlorophenyl)ethanamine

Appearance: Clear yellow Liquid

Kaava: C₈H₁₀ClN
Kiehumispiste: 232 °C (760 mmHg)
Tiheys: 1,1 g/cm³
Leimahduspiste: 103 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00044766
CAS-numero: 6299-02-1
UN: 2810
ADR: 6.1,II

Order Now

Testitulosten erittely

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5455 to 1.5475 (20°C, 589 nm)
GC ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR