1,1,2,2-Tetrakloorietaani-D2 (99.5% D) for NMR spectroscopy

Toimittaja: Thermo Fisher Scientific
Vaara

321310010 321310050 321310250
ACRO321310010EA 28.2 EUR
ACRO321310010 ACRO321310050 ACRO321310250
1,1,2,2-Tetrakloorietaani-D2 (99.5% D) for NMR spectroscopy
1,1,2,2-Tetrakloorietaani-D2
Kaava: Cl₂CDCDCl₂
Kiehumispiste: 146 °C (760 mmHg)
Sulamispiste: -43 °C
Tiheys: 1,61 g/cm³
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00037672
CAS-numero: 33685-54-0
EINECS: 251-634-1
UN: 1702
ADR: 6.1,II

Order Now

Testitulosten erittely

Appearance Clear colorless Liquid
Water ≤0.03 %
D-Enrichment ≥99.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR