Asetoni-D6 100.0 atom % D for NMR spectroscopy

Toimittaja: Thermo Fisher Scientific
Vaara

320610075
ACRO320610075EA 130 EUR
ACRO320610075
Asetoni-D6 100.0 atom % D for NMR spectroscopy
Asetoni-D6

Packaged in 0.75ml ampoules

Kaava: D₃CC(O)CD₃
Kiehumispiste: 55,5 °C (1013 hPa)
Sulamispiste: –93 °C
Tiheys: 0,87 g/cm³
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00044635
CAS-numero: 666-52-4
EINECS: 211-563-9
UN: 1090
ADR: 3,II

Order Now

Testitulosten erittely

Appearance Clear colorless Liquid
Water ≤0.02 % (Coulometric)
D-Enrichment ≥99.95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR