Osmium(III) chloride hydrate

Toimittaja: Thermo Fisher Scientific
Vaara

318705000 318700025
ACRO318705000EA 187 EUR
ACRO318705000 ACRO318700025
Osmium(III) chloride hydrate
Osmium(III) chloride hydrate
Kaava: Cl₃Os·H₂O
Sulamispiste: >500 °C (haj. lämpötila)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149815
CAS-numero: 14996-60-2
EINECS: 236-587-7
UN: 2923
ADR: 8,III

Order Now

Testitulosten erittely

Appearance Dark red to black Powder or crystals
Osmium (Os) 53 to 56 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR