Triammoniumdodekamolybdofosfaatti-H2O

Toimittaja: Thermo Fisher Scientific
Varoitus

317100500 317100010 317102500
ACRO317100500EA 87.1 EUR
ACRO317100500 ACRO317100010 ACRO317102500
Triammoniumdodekamolybdofosfaatti-H2O
Triammoniumdodekamolybdofosfaatti-H2O
Kaava: H₁₂Mo₁₂N₃O₄₀P·H₂O
Storage Temperature: Jääkaappi
MDL numero: MFCD00150423
CAS-numero: 54723-94-3
EINECS: 234-714-0
ADR: 8,III

Order Now

Testitulosten erittely

Appearance Yellow Powder
Chloride (Cl) ≤500 ppm
Sulfate (SO4) ≤250 ppm
Calcium (Ca) ≤200 ppm
Iron (Fe) ≤50 ppm
Lead (Pb) ≤50 ppm
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR