Chloropentacarbonylrhenium 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: Pentacarbonylchlororhenium(I), Chloropentacarbonylrhenium(I), Rhenium pentacarbonyl chloride

316310010 316310050
ACRO316310010EA 209 EUR
ACRO316310010 ACRO316310050
Chloropentacarbonylrhenium 98%
Chloropentacarbonylrhenium
Kaava: C₅ClO₅Re
MW: 361.71 g/mol
MDL numero: MFCD00013296
CAS-numero: 14099-01-5
EINECS: 237-948-1
UN: 2811
ADR: 6.1 III

Order Now

Testitulosten erittely

Appearance White to off-white Powder
Elemental Analysis ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR