Rauta(II)sulfidi 99.9% (trace metals basis) -100 mesh

Toimittaja: Thermo Fisher Scientific

315870100 315870500
ACRO315870100EA 97.6 EUR
ACRO315870100 ACRO315870500
Rauta(II)sulfidi 99.9% (trace metals basis) -100 mesh
Rauta(II)sulfidi
Kaava: FeS
Sulamispiste: 1195 °C
Tiheys: 4,84 g/cm³
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011013
CAS-numero: 1317-37-9
EINECS: 215-268-6
ADR: 9,III
Merck Index: 12,04106

Order Now

Testitulosten erittely

Appearance Grey to brown to black Powder
Total trace metal impurities ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR