(1S,2R)-1-Amino-2,3-dihydro-1H-inden-2-ol 99%

Toimittaja: Thermo Fisher Scientific
Varoitus

315652500 315650010 315650050
ACRO315652500EA 24 EUR
ACRO315652500 ACRO315650010 ACRO315650050
(1S,2R)-1-Amino-2,3-dihydro-1H-inden-2-ol 99%
(1S,2R)-1-Amino-2,3-dihydro-1H-inden-2-ol

Appearance: White to beige Powder

Kaava: C₉H₁₁NO
Sulamispiste: 117…121 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00216655
CAS-numero: 126456-43-7

Order Now

Testitulosten erittely

Appearance White to beige Powder
Infrared spectrum Conforms
Melting point 117 to 121 °C
GC ≥98.5 %
Specific optical rotation -41.5° to -46.5° (20 °C, 589 nm) (c=1, CH3OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR