N-Fluorobenzenesulphonimide 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

311900050 311900250
ACRO311900050EA 81.5 EUR
ACRO311900050 ACRO311900250
N-Fluorobenzenesulphonimide 97%
N-Fluorobenzenesulphonimide
Kaava: C₁₂H₁₀FNO₄S₂
Sulamispiste: 110…115 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00144885
CAS-numero: 133745-75-2

Order Now

Testitulosten erittely

Appearance Off-white to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 110 to 117 °C
Titration with Na2S2O3 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR