2-Tolylboronic acid 95%

Toimittaja: Thermo Fisher Scientific
Varoitus

309260010 309260050
ACRO309260010EA 37.1 EUR
ACRO309260010 ACRO309260050
2-Tolylboronic acid 95%
2-Tolylboronic acid
Kaava: C₇H₉BO₂
Sulamispiste: 164…165 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00093526
CAS-numero: 16419-60-6

Order Now

Testitulosten erittely

Appearance White to beige Powder or crystals
Infrared spectrum Conforms
Melting point 160 to 166 °C
Additional info May contain varying amounts of anhydrides
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR