3,5-Bis(trifluorimetyyli)bentsyylibromidi 97%

Toimittaja: Thermo Fisher Scientific
Vaara

303640010 303640050
ACRO303640010EA 45.6 EUR
ACRO303640010 ACRO303640050
3,5-Bis(trifluorimetyyli)bentsyylibromidi 97%
3,5-Bis(trifluorimetyyli)bentsyylibromidi
Kaava: C₉H₅BrF₆
Kiehumispiste: 136…140 °C (14 mmHg)
Tiheys: 1,67 g/cm³
Leimahduspiste: 26 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00009905
CAS-numero: 32247-96-4
EINECS: 250-971-1
UN: 2920
ADR: 8,II

Order Now

Testitulosten erittely

Appearance Clear colorless to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4460 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR