L-Proline methyl ester hydrochloride 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

303000050 303000010 303000250
ACRO303000050EA 97.7 EUR
ACRO303000050 ACRO303000010 ACRO303000250
L-Proline methyl ester hydrochloride 98%
L-Proline methyl ester hydrochloride

CAS No.: 2133-40-6

Kaava: C₆H₁₁NO₂·HCl
Sulamispiste: 71 °C
Storage Temperature: Pakastin
MDL numero: MFCD00012708
CAS-numero: 2133-40-6

Order Now

Testitulosten erittely

Appearance White Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
Water ≤0.5 % (K.F.)
Specific optical rotation -31° to -35° (20°C, 589 nm) (c=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR