D-(+)-Arabitol 99%

Toimittaja: Thermo Fisher Scientific

302870050 302870250 302871000
ACRO302870050EA 59.7 EUR
ACRO302870050 ACRO302870250 ACRO302871000
D-(+)-Arabitol 99%
D-(+)-Arabitol
Kaava: C₅H₁₂O₅
Sulamispiste: 97…105 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00004709
CAS-numero: 488-82-4
EINECS: 207-686-2

Order Now

Testitulosten erittely

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 97 to 105 °C
HPLC ≥98.5 % (ELSD)
Specific optical rotation +10.5° to +13.5° (20 °C, 589 nm) (c=2, 2 % borax)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR