(±)-α-Terpineol 97+%

Toimittaja: Thermo Fisher Scientific
Varoitus

301610250 301615000 301610025
ACRO301610250EA 33.6 EUR
ACRO301610250 ACRO301615000 ACRO301610025
(±)-α-Terpineol 97+%
(±)-α-Terpineol
Kaava: C₁₀H₁₈O
Kiehumispiste: 214…224 °C (760 mmHg)
Sulamispiste: 31…34 °C
Tiheys: 0,937 g/cm³
Leimahduspiste: 90 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00001557
CAS-numero: 98-55-5
EINECS: 202-680-6

Order Now

Testitulosten erittely

Appearance Clear colorless to white solid
Infrared spectrum Conforms
Refractive index 1.4800 to 1.4850 (20°C, 589 nm)
GC ≥97.0 %
Specific gravity (20°C) 0.934 to 0.940


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR