1-Aminocyclopropanecarboxylic acid 99%

Toimittaja: Thermo Fisher Scientific
Varoitus

301422500 301420010
ACRO301422500EA 191 EUR
ACRO301422500 ACRO301420010
1-Aminocyclopropanecarboxylic acid 99%
1-Aminocyclopropanecarboxylic acid
Kaava: C₄H₇NO₂
Sulamispiste: 257 °C (haj. lämpötila)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00009944
CAS-numero: 22059-21-8

Order Now

Testitulosten erittely

Appearance White to off-white Powder and/or granules
Infrared spectrum Conforms
Water ≤9 % (K.F.)
Titration with NaOH ≥98.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR