2-Phenylquinoline 99+%

Toimittaja: Thermo Fisher Scientific
Varoitus

297990010 297990050
ACRO297990010EA 92.2 EUR
ACRO297990010 ACRO297990050
2-Phenylquinoline 99+%
2-Phenylquinoline
Kaava: C₁₅H₁₁N
Kiehumispiste: 363 °C (760 mmHg)
Sulamispiste: 82…84 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011568
CAS-numero: 612-96-4
EINECS: 210-326-7

Order Now

Testitulosten erittely

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 82 to 84 °C
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR