L(+)- Litiumlaktaatti 99%, pure

Toimittaja: Thermo Fisher Scientific

297610250 297610050 297611000 297615000
ACRO297610250EA 52.4 EUR
ACRO297610250 ACRO297610050 ACRO297611000 ACRO297615000
L(+)- Litiumlaktaatti 99%, pure
L(+)- Litiumlaktaatti
Kaava: C₃H₅LiO₃
Kiehumispiste: 200 °C
Sulamispiste: >75 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00065512
CAS-numero: 27848-80-2
EINECS: 248-692-5

Order Now

Testitulosten erittely

Appearance White Crystalline powder
Infrared spectrum Conforms
Specific optical rotation -13° to -16° (20°C, 589 nm) (c=1, H2O)
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR