Litiumasetaatti, vedetön 99+% for analysis

Toimittaja: Thermo Fisher Scientific
Varoitus

297110250 297111000
ACRO297110250EA 61.9 EUR
ACRO297110250 ACRO297111000
Litiumasetaatti, vedetön 99+% for analysis
Litiumasetaatti
Kaava: H₃CCOOLi
Sulamispiste: 280…285 °C
Tiheys: 1,26 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00013057
CAS-numero: 546-89-4
EINECS: 208-914-3
Merck Index: 12,05544

Order Now

Testitulosten erittely

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 280 to 285 °C
Water ≤0.3 % (K.F.)
Titration with HClO4 ≥99.0 %
Chloride (Cl) ≤200 ppm
Sulfate (SO4) ≤200 ppm
Calcium (Ca) ≤200 ppm
Iron (Fe) ≤10 ppm
Magnesium (Mg) ≤10 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤100 ppm
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR