(2-Bromifenyyli)etikkahappo 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

296650100 296650050
ACRO296650100EA 35.9 EUR
ACRO296650100 ACRO296650050
(2-Bromifenyyli)etikkahappo 98%
(2-Bromifenyyli)etikkahappo
Kaava: (C₆H₄Br)CH₂COOH
Kiehumispiste: 308 °C (1013 hPa)
Sulamispiste: 104…106 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00004314
CAS-numero: 18698-97-0
EINECS: 242-509-2

Order Now

Testitulosten erittely

Appearance White to light beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 105 to 110 °C
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR