Syklopenteeni 95+%

Toimittaja: Thermo Fisher Scientific
Vaara

295905000 295900025 295901000
ACRO295905000EA 173 EUR
ACRO295905000 ACRO295900025 ACRO295901000
Syklopenteeni 95+%
Syklopenteeni
Kaava: C₅H₈
MW: 68.12 g/mol
Kiehumispiste: 44 °C (760 mmHg)
Sulamispiste: –135 °C
Tiheys: 0.774 g/cm³
Leimahduspiste: –30 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00001394
CAS-numero: 142-29-0
EINECS: 205-532-9
UN: 2246
ADR: 3 II

Order Now

Testitulosten erittely

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4200 to 1.4240 (20°C, 589 nm)
GC ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR