Terbium trinitrate hexahydrate 99.99+% (trace metals basis)

Toimittaja: Thermo Fisher Scientific
Vaara

295790020 295790100
ACRO295790020EA 64.6 EUR
ACRO295790020 ACRO295790100
Terbium trinitrate hexahydrate 99.99+% (trace metals basis)
Terbium trinitrate hexahydrate
Kaava: N₃O₉Tb·6H₂O
Sulamispiste: 89,3 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149868
CAS-numero: 13451-19-9
EINECS: 233-138-7
UN: 3077
ADR: 9,III

Order Now

Testitulosten erittely

Appearance White Granular powder
Assay ≥99.99 % (TREO base)
Total conc. of imp.as REO ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR