1-Cyclohexene-1-carboxylic acid 96%

Toimittaja: Thermo Fisher Scientific
Varoitus

291750025
ACRO291750025EA 104 EUR
ACRO291750025
1-Cyclohexene-1-carboxylic acid 96%
1-Cyclohexene-1-carboxylic acid
Kaava: C₇H₁₀O₂
Kiehumispiste: 137 °C (15 mmHg)
Sulamispiste: 35…39 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00001545
CAS-numero: 636-82-8
ADR: 8,III

Order Now

Testitulosten erittely

Appearance White to light yellow Crystalline mass or liquid
Infrared spectrum Conforms
HPLC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR