(Aminomethyl)phosphonic acid 99%

Toimittaja: Thermo Fisher Scientific
Vaara

291640010 291642500 291640050
ACRO291640010EA 165 EUR
ACRO291640010 ACRO291642500 ACRO291640050
(Aminomethyl)phosphonic acid 99%
(Aminomethyl)phosphonic acid
Kaava: CH₆NO₃P
Sulamispiste: >300 °C (haj. lämpötila)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008105
CAS-numero: 1066-51-9
UN: 3261
ADR: 8,III

Order Now

Testitulosten erittely

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR