N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate) 96%

Toimittaja: Thermo Fisher Scientific
Vaara

291490050 291490250 291491000
ACRO291490050EA 44.9 EUR
ACRO291490050 ACRO291490250 ACRO291491000
N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate) 96%
N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate)

CAS No.: 140681-55-6

Kaava: C₇H₁₄B₂ClF₉N₂
Sulamispiste: 260 °C (haj. lämpötila)
Leimahduspiste: >54 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00142607
CAS-numero: 140681-55-6
UN: 3088
ADR: 4.2,II

Order Now

Testitulosten erittely

Appearance White Powder
Infrared spectrum Conforms
Titration Iodometric ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR