Ruthenium(III) chloride oxide ≥85%, pure

Toimittaja: Thermo Fisher Scientific
Varoitus

279710010 279710050
ACRO279710010EA 141 EUR
ACRO279710010 ACRO279710050
Ruthenium(III) chloride oxide ≥85%, pure
Ruthenium(III) chloride oxide
Kaava: H₄₂Cl₆N₁₄O₂Ru₃
Sulamispiste: >500 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00011479
CAS-numero: 11103-72-3

Order Now

Testitulosten erittely

Appearance Dark red or purple Powder
Elemental Analysis 34 to 39 % (Ru)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR