Polyvinyylipyrrolidoni, M.W. ∼ 3500 (K12)

Toimittaja: Thermo Fisher Scientific

Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP

276142500 276140010
ACRO276142500EA 65.5 EUR
ACRO276142500 ACRO276140010
Polyvinyylipyrrolidoni, M.W. ∼ 3500 (K12)
Polyvinyylipyrrolidoni
Kaava: (C₆H₉NO)n
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01076626
CAS-numero: 9003-39-8

Order Now

Testitulosten erittely

Appearance White to yellowish-white Powder or flakes
Infrared spectrum Conforms
Water ≤5.0 % (K.F.)
Sulfated ash ≤0.1 %
Additional info European color test - R color: R6
HPLC ≤3.0 % 2-Pyrrolidone
Identification Passes test
pH 3.0 to 5.0 (5 % in water)
Nitrogen (N) 11.5 to 12.8 % (on anhydrous substance)
Heavy metals (as Pb) ≤10 ppm
Aldehyde ≤0.05 %
Hydrazine (N2H4) ≤1 ppm
Peroxides ≤400 ppm (Titanyl sulphate method)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR