N-Carbobenzyloxy-L-valine 99+%

Toimittaja: Thermo Fisher Scientific

275940050 275940250
ACRO275940050EA 35.2 EUR
ACRO275940050 ACRO275940250
N-Carbobenzyloxy-L-valine 99+%
N-Carbobenzyloxy-L-valine

Appearance: White to off-white Crystalline powder

Kaava: C₁₃H₁₇NO₄
Sulamispiste: 58…60 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008922
CAS-numero: 1149-26-4

Order Now

Testitulosten erittely

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Sulfated ash ≤0.2 %
HPLC ≥98.0 %
Specific optical rotation -3.2° to -4.8° (20°C, 589 nm) (c=2, acetic acid)
Titration with NaOH ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR