2,2'-Oxybis(ethylamine) 98%

Toimittaja: Thermo Fisher Scientific
Vaara

275840010 275840050
ACRO275840010EA 127 EUR
ACRO275840010 ACRO275840050
2,2'-Oxybis(ethylamine) 98%
2,2'-Oxybis(ethylamine)

CAS No.: 2752-17-2

Kaava: C₄H₁₂N₂O
Kiehumispiste: 69…71 °C (15 mmHg)
Tiheys: 0,961 g/cm³
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00466591
CAS-numero: 2752-17-2
UN: 2735
ADR: 8,III

Order Now

Testitulosten erittely

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4550 to 1.4580 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR