N(α)-Boc-L-tryptophan 97%

Toimittaja: Thermo Fisher Scientific

275690050 275690250 275691000
ACRO275690050EA 31.6 EUR
ACRO275690050 ACRO275690250 ACRO275691000
N(α)-Boc-L-tryptophan 97%
N(α)-Boc-L-tryptophan
Kaava: C₁₆H₂₀N₂O₄
Sulamispiste: 134…139 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00065595
CAS-numero: 13139-14-5
EINECS: 236-072-7

Order Now

Testitulosten erittely

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤1.0 % (K.F.)
HPLC ≥96.0 %
Specific optical rotation -19° to -23° (20°C, 589 nm) (c=1, CH3COOH)
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR