N(α)-Boc-L-histidine 99+%

Toimittaja: Thermo Fisher Scientific

275590010 275590050 275590250
ACRO275590010EA 29.7 EUR
ACRO275590010 ACRO275590050 ACRO275590250
N(α)-Boc-L-histidine 99+%
N(α)-Boc-L-histidine
Kaava: C₁₁H₁₇N₃O₄
Sulamispiste: 196…199 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00065576
CAS-numero: 17791-52-5
EINECS: 241-768-9

Order Now

Testitulosten erittely

Appearance White to off-white Powder or crystals and/or chunks
Infrared spectrum Conforms
TLC ≥99.0 %
Specific optical rotation +24.5° to +26.5° (20°C, 589 nm) (c=1, CH3OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR