Dibenzyl azodicarboxylate 94%

Toimittaja: Thermo Fisher Scientific
Varoitus

270270250 270270050
ACRO270270250EA 215 EUR
ACRO270270250 ACRO270270050
Dibenzyl azodicarboxylate 94%
Dibenzyl azodicarboxylate
Kaava: C₁₆H₁₄N₂O₄
Sulamispiste: 44…52 °C
Leimahduspiste: >230 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00016737
CAS-numero: 2449-05-0
EINECS: 219-508-0
UN: 1325
ADR: 4.1,III

Order Now

Testitulosten erittely

Appearance Yellow to orange Crystalline powder
Infrared spectrum Conforms
Melting point 44 to 52 °C
Loss on drying ≤0.75 %
UV spectrophotometry ≥92 %
Heavy metals (as Pb) ≤30 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR